Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Copper(II) nitrate hydrate, Puratronic™, 99.999% (metals basis)

CAS: 13778-31-9 | CuN2O6 | 187.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Catalog number 010699.36
also known as 010699-36
Price (EUR)
809,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
809,00
Each
Chemical Identifiers
CAS13778-31-9
IUPAC Namecopper(2+) dinitrate
Molecular FormulaCuN2O6
InChI KeyXTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It serves as a catalyst for oxidative coupling of 2,6-dimethylphenol, resulting in an important engineering polymer material. Also used in textiles and polishing agents for other metals. It is a precursor for copper oxide, which finds use as a catalyst for many organic transformations. Menke conditions employ copper nitrate for aromatic nitrations. Claycop reagent, the hydrated copper nitrate deposited on clay, finds use in many organic reactions, for example, oxidation of dithioacetals to carbonyls and thiols to disulfides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only