Boron carbide, 90+%
Boron carbide, 90+%
Boron carbide, 90+%
Thermo Scientific Chemicals

Boron carbide, 90+%

CAS: 12069-32-8 | CB4 | 55.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010922.22
also known as 010922-22
Price (EUR)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS12069-32-8
IUPAC Name2,3,4,5-tetraboratetracyclo[2.1.0.0¹,³.0²,⁵]pentane
Molecular FormulaCB4
InChI KeyINAHAJYZKVIDIZ-UHFFFAOYSA-N
SMILESB12B3B4B1C234
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)90.0% min
Boron carbide is a hard boron-carbon ceramic material used in tank armor, bulletproof vests, engine sabotage powders, neutron absorber, cutting tools and dies and in brake linings of vehicles. It acts as an antioxidant additives in magnesia-carbon bricks. It is used as a precursor in the production of boron containing materials such as titanium boride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only