Calcium oxide, 99.95% (metals basis)
Calcium oxide, 99.95% (metals basis)
Calcium oxide, 99.95% (metals basis)
Thermo Scientific Chemicals

Calcium oxide, 99.95% (metals basis)

CAS: 1305-78-8 | CaO | 56.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 010923.18
also known as 010923-18
Price (EUR)
73,90
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
73,90
Each
Chemical Identifiers
CAS1305-78-8
IUPAC Namecalcium oxidandiide
Molecular FormulaCaO
InChI KeyBRPQOXSCLDDYGP-UHFFFAOYSA-N
SMILES[O--].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
CommentMay contain traces of Ca(OH)2 which disintegrates upon heating up to 120?
Total Metal Impurities0.05% max.
Particle Size-400 Mesh
It is used in cement, glass, steel, paper, chemical and petroleum industries. It acts as a key ingredient in the process of cement making. It is widely used industrially as a dehydrating agent and an absorbent. Further it finds use as a water softener and as a pH regulator for wastewater. It is useful to detect traces of water in hydrocarbons, and in flue-gas desulfurization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only