Copper foil, 1.0mm (0.04in) thick, 99.99% (metals basis)
Copper foil, 1.0mm (0.04in) thick, 99.99% (metals basis)
Copper foil, 1.0mm (0.04in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals

Copper foil, 1.0mm (0.04in) thick, 99.99% (metals basis)

Form: approx. 2.2g/50x50mm | CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
50 x 250 mm
50 x 1000 mm
Catalog number 010954.HC
also known as 010954-HC
Price (EUR)
384,65
Online Exclusive
427,00
Save 42,35 (10%)
Each
Quantity:
50 x 1000 mm
Request bulk or custom format
Price (EUR)
384,65
Online Exclusive
427,00
Save 42,35 (10%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil
Appearance (Color)Copper
Comment0.9 - 1.1mm thick
Total Metal Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only