Antimony shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9999% (metals basis)
Antimony shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9999% (metals basis)
Antimony shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9999% (metals basis)
Thermo Scientific Chemicals

Antimony shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9999% (metals basis)

CAS: 7440-36-0 | Sb | 121.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 011071.18
also known as 011071-18
Price (EUR)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS7440-36-0
IUPAC Nameantimony
Molecular FormulaSb
InChI KeyWATWJIUSRGPENY-UHFFFAOYSA-N
SMILES[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only