Aluminum phosphate, 97%
Aluminum phosphate, 97%
Aluminum phosphate, 97%
Thermo Scientific Chemicals

Aluminum phosphate, 97%

CAS: 7784-30-7 | AlO4P | 121.951 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number 011109.30
also known as 011109-30
Price (EUR)
58,90
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
58,90
Each
Chemical Identifiers
CAS7784-30-7
IUPAC Namealuminium(3+) phosphate
Molecular FormulaAlO4P
InChI KeyILRRQNADMUWWFW-UHFFFAOYSA-K
SMILES[Al+3].[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Assay (unspecified)Al2O3: 39-42%
Assay (unspecified)P2O5: 58-62%
As a flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy.Aluminum phosphate is used as catalysts, molecular sieves, flux for ceramics and as a gel in pharmacy. It is employed in the preparation of dental cement, emollients and fire-resistant coating. It plays an important role as an anti-fouling agent in textile industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only