Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Germanium(IV) oxide, Puratronic™, 99.999% (metals basis)

CAS: 1310-53-8 | GeO2 | 104.628 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 011155.06
also known as 011155-06
Price (EUR)
79,60
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
79,60
Each
Chemical Identifiers
CAS1310-53-8
IUPAC Namegermanedione
Molecular FormulaGeO2
InChI KeyYBMRDBCBODYGJE-UHFFFAOYSA-N
SMILESO=[Ge]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
Germanium dioxide is used as an optical material for wide-angle lenses and in optical microscope objective lenses. It is used as a catalyst in the preparation of polyethylene terephthalate resin. Further, it is used as a feedstock for the manufacturing of phosphors and semiconductor materials. It is also used in transistors and diodes, and infrared transmitting glass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only