Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Terbium(III) chloride hexahydrate, REacton™, 99.99% (REO)

CAS: 13798-24-8 | Cl3H12O6Tb | 373.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011210.09
also known as 011210-09
Price (EUR)
155,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
155,00
Each
Chemical Identifiers
CAS13798-24-8
IUPAC Nameterbium(3+) hexahydrate trichloride
Molecular FormulaCl3H12O6Tb
InChI KeyULJUVCOAZNLCJZ-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Terbium(III) chloride hexahydrate is used in the primary chemical intermediates and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only