Holmium(III) oxide, REacton™, 99.99% (REO)
Holmium(III) oxide, REacton™, 99.99% (REO)
Holmium(III) oxide, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Holmium(III) oxide, REacton™, 99.99% (REO)

CAS: 12055-62-8 | Ho2O3 | 377.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 011281.09
also known as 011281-09
Price (EUR)
165,00
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
165,00
Each
Chemical Identifiers
CAS12055-62-8
IUPAC Namediholmium(3+) trioxidandiide
Molecular FormulaHo2O3
InChI KeyOWCYYNSBGXMRQN-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Ho+3].[Ho+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Holmium(III) oxide iacts as colorants which is used for cubic zirconia and glass, as a calibration standard for optical spectrophotometers, as a specialty catalyst, phosphor and a laser material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only