Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Gadolinium(III) chloride hexahydrate, REacton™, 99.99% (REO)

CAS: 13450-84-5 | Cl3GdH12O6 | 371.69 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 011287.06
also known as 011287-06
Price (EUR)
97,80
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
97,80
Each
Chemical Identifiers
CAS13450-84-5
IUPAC Namegadolinium(3+) hexahydrate trichloride
Molecular FormulaCl3GdH12O6
InChI KeyPNYPSKHTTCTAMD-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Gd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Gadolinium(III) chloride hexahydrate is employed in synthesis and NMR studies of the paramagnetic GdPCTA-[12] and GdPCTA-[13] complexes with possible use as contrast agents for magnetic resonance imaging..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only