Silver foil, 2.0mm (0.08in) thick, hard, 99.9% (metals basis)
Silver foil, 2.0mm (0.08in) thick, hard, 99.9% (metals basis)
Silver foil, 2.0mm (0.08in) thick, hard, 99.9% (metals basis)
Thermo Scientific Chemicals

Silver foil, 2.0mm (0.08in) thick, hard, 99.9% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 011437.FF
also known as 011437-FF
Price (EUR)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
FormFoil, 2.0mm (0.08in) thick.
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only