Rhodium foil, 0.025mm (0.001in) thick, 99.8% (metals basis)
Rhodium foil, 0.025mm (0.001in) thick, 99.8% (metals basis)
Rhodium foil, 0.025mm (0.001in) thick, 99.8% (metals basis)
Thermo Scientific Chemicals

Rhodium foil, 0.025mm (0.001in) thick, 99.8% (metals basis)

CAS: 7440-16-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 011524.FF
also known as 011524-FF
Price (EUR)
-
Quantity:
25 x 25 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-16-6
IUPAC Namerhodium
Molecular FormulaRh
InChI KeyMHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES[Rh]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.025 mm (0.001 in) thick
Total Metal Impurities≤0.2%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.