Sodium acetate trihydrate, ACS, 99.0%-101%
Sodium acetate trihydrate, ACS, 99.0%-101%
Sodium acetate trihydrate, ACS, 99.0%-101%
Thermo Scientific Chemicals

Sodium acetate trihydrate, ACS, 99.0%-101%

CAS: 6131-90-4 | C2H9NaO5 | 136.079 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 011553.A3
also known as 011553-A3
Price (EUR)
98,65
キャンペーン価格
109,00
Save 10,35 (9%)
Each
Quantity:
2 kg
Request bulk or custom format
Price (EUR)
98,65
キャンペーン価格
109,00
Save 10,35 (9%)
Each
Chemical Identifiers
CAS6131-90-4
IUPAC Namesodium acetate trihydrate
Molecular FormulaC2H9NaO5
InChI KeyAYRVGWHSXIMRAB-UHFFFAOYSA-M
SMILESO.O.O.[Na+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentSubstances reducing permanganate P.T.
Insoluble material0.005%
pHof a 5% solution 7.5-9.2 at 25?
Assay (unspecified)99.0-101% NaC₂H₃O₂?3H₂O
Impurity contentCa 0.005%
View more
In photography, analytical laboratory as buffer and in pharmaceutical.Sodium acetate trihydrate is used in various applications. It is used as a buffer of a pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as a mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid, in the textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only