Zinc acetate dihydrate, ACS, 98.0-101.0%
Zinc acetate dihydrate, ACS, 98.0-101.0%
Zinc acetate dihydrate, ACS, 98.0-101.0%
Zinc acetate dihydrate, ACS, 98.0-101.0%
Thermo Scientific Chemicals

Zinc acetate dihydrate, ACS, 98.0-101.0%

CAS: 5970-45-6 | C4H10O6Zn | 219.498 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 011559.A1
also known as 011559-A1
Price (EUR)
74,97
Online Exclusive
88,20
Save 13,23 (15%)
Each
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
74,97
Online Exclusive
88,20
Save 13,23 (15%)
Each
Add to cart
Chemical Identifiers
CAS5970-45-6
IUPAC Namezinc(2+) diacetate dihydrate
Molecular FormulaC4H10O6Zn
InChI KeyBEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILESO.O.[Zn++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or solid
Complexometric Titration98.0-101.0%
Calcium content≤0.005%
Chloride content≤5ppm
View more
Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only