Potassium iodide, ACS, 99.0% min
Potassium iodide, ACS, 99.0% min
Potassium iodide, ACS, 99.0% min
Thermo Scientific Chemicals

Potassium iodide, ACS, 99.0% min

For Use in Photographic Emulsions, in Analytical Chemistry, As a Feed Additive | CAS: 7681-11-0 | IK | 166.003 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Catalog number 011601.22
also known as 011601-22
Price (EUR)
55,10
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
55,10
Each
Chemical Identifiers
CAS7681-11-0
IUPAC Namepotassium iodide
Molecular FormulaIK
InChI KeyNLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES[K+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentIO₃ 3ppm
pHof a 5% solution 6.0-9.2 at 25?
Assay (unspecified)99.0% KI min
Impurity contentCa 0.002%
Impurity contentFe 3ppm
View more
It is both a source of iodide and a mild reducing agent. It is used as a component in the electrolyte of dye sensitized solar cells. In the field of synthetic organic chemistry, it is used in the preparation of alkyl or aryl iodo compounds. It is employed as a nucleophilic catalyst in the alkylations with alkyl bromide, chloride, tosylate and mesylate. In the presence of acid it forms hydroiodic acid, which is a strong reducing agent. It is useful in producing Lugol’s solution (or Lugol’s iodine) which finds many applications including starch testing, gram stains, and etching of gold surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only