Iron(II) perchlorate hydrate, Reagent Grade
Iron(II) perchlorate hydrate, Reagent Grade
Iron(II) perchlorate hydrate, Reagent Grade
Iron(II) perchlorate hydrate, Reagent Grade
Thermo Scientific Chemicals

Iron(II) perchlorate hydrate, Reagent Grade

CAS: 335159-18-7 | Cl2FeO8 | 254.74 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 011646.22
also known as 011646-22
Price (EUR)
46,60
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
46,60
Each
Chemical Identifiers
CAS335159-18-7
IUPAC Nameλ²-iron(2+) diperchlorate
Molecular FormulaCl2FeO8
InChI KeyCKFMJXZQTNRXGX-UHFFFAOYSA-L
SMILES[Fe++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)14.5-16.0% Fe²⁺
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only