Barium perchlorate, anhydrous
Barium perchlorate, anhydrous
Barium perchlorate, anhydrous
Thermo Scientific Chemicals

Barium perchlorate, anhydrous

CAS: 13465-95-7 | BaCl2O8 | 336.219 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 011657.22
also known as 011657-22
Price (EUR)
64,00
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
64,00
Each
Chemical Identifiers
CAS13465-95-7
IUPAC Namebarium(2+) diperchlorate
Molecular FormulaBaCl2O8
InChI KeyOOULUYZFLXDWDQ-UHFFFAOYSA-L
SMILES[Ba++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting Point503.0-506.0°C
FormPowder or crystals or crystalline powder or granules
Assay from Suppliers CofAReport Value
Barium perchlorate, anhydrous is used to make explosives. It is also used as desiccant, ribonuclease determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only