Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min
Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min
Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min
Thermo Scientific Chemicals

Potassium hexacyanoruthenate(II) hydrate, Ru 23.0% min

CAS: 339268-21-2 | C6K4N6Ru | 413.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.2 g
2 g
10 g
Catalog number 011876.SS
also known as 011876-SS
Price (EUR)
67,50
Each
Quantity:
0.2 g
Request bulk or custom format
Price (EUR)
67,50
Each
Chemical Identifiers
CAS339268-21-2
IUPAC Nametetrapotassium hexacyanorutheniumbis(ylium)
Molecular FormulaC6K4N6Ru
InChI KeyFULSJSKUKUZHLZ-UHFFFAOYSA-N
SMILES[K+].[K+].[K+].[K+].N#C[Ru++](C#N)(C#N)(C#N)(C#N)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ru 23.0% min
Potassium hexacyanoruthenate(II) hydrate used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only