Ethylenediaminetetraacetic acid, ACS, 99.4+%
Ethylenediaminetetraacetic acid, ACS, 99.4+%
Ethylenediaminetetraacetic acid, ACS, 99.4+%
Thermo Scientific Chemicals

Ethylenediaminetetraacetic acid, ACS, 99.4+%

Chelating agent | CAS: 60-00-4 | C10H16N2O8 | 292.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 011931.36
also known as 011931-36
Price (EUR)
101,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
101,00
Each
Chemical Identifiers
CAS60-00-4
IUPAC Name2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
Molecular FormulaC10H16N2O8
InChI KeyKCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILESOC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline powder or powder or solid
Complexometric Titration99.4-100.6%
Calcium content≤0.001%
Iron content≤0.005%
Residue on ignition≤0.2%
View more
Chelating agentEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and is used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only