Platinum foil, 1.0mm (0.04in) thick, Premion™, 99.997% (metals basis)
Platinum foil, 1.0mm (0.04in) thick, Premion™, 99.997% (metals basis)
Platinum foil, 1.0mm (0.04in) thick, Premion™, 99.997% (metals basis)
Thermo Scientific Chemicals

Platinum foil, 1.0mm (0.04in) thick, Premion™, 99.997% (metals basis)

CAS: 7440-06-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 012060.FF
also known as 012060-FF
Price (EUR)
-
Quantity:
25 x 25 mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS6-4-7440
Percent Purity99.997%
Quantity25 x 25 mm
Unit SizeEach