Spectroflux 100, Lithium tetraborate
Spectroflux 100, Lithium tetraborate
Thermo Scientific Chemicals

Spectroflux 100, Lithium tetraborate

CAS: 12007-60-2
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 x 1 kg
Catalog number 012078.D9
also known as 012078-D9
Price (EUR)
-
Quantity:
5 x 1 kg
Request bulk or custom format
Chemical Identifiers
CAS12007-60-2
IUPAC Namedilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Li2O7
InChI KeyRDZCISUGRQUANA-UHFFFAOYSA-N
SMILES[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormPowder
Elemental AnalysisBoron, B (theoretical: 25.56%): 25.40-25.60%
Elemental AnalysisLithium, Li (theoretical: 8.21%): 8.20-8.30%
Impurity contentSilicon, Si : ≤50ppm
View more
A general purpose acidic flux for fusing basic oxides, carbonates, alumino-silicates, glass, ceramics, cement steel, aluminum, concrete mixes, soils, magnesites, bauxites and rare earth oxides. Dissolves most refractories. Not suitable for highly acidic samples. Samples must be fully oxidized.Spectroflux 100, Lithium tetraborate is used in enamels. Further, it is a spectroscopic fluxing agent which is utilized for fusing basic oxides, glass, alumino-silicates, ceramics, carbonates, cement, steel, aluminum, soil, concrete mixes, bauxites, magnesites and rare earth oxides. In addition to this, it is used for dissolution of samples containing a high concentration of basic oxides and carbonate rocks and aluminosilicates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A general purpose acidic flux for fusing basic oxides, carbonates, alumino-silicates, glass, ceramics, cement steel, aluminum, concrete mixes, soils, magnesites, bauxites and rare earth oxides. Dissolves most refractories. Not suitable for highly acidic samples. Samples must be fully oxidized.