Copper(I) cyanide
Copper(I) cyanide
Copper(I) cyanide
Thermo Scientific Chemicals

Copper(I) cyanide

For Electroplating Cu or Fe, Polymerization Catalyst | CAS: 544-92-3 | CCuN | 89.564 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2 x 2.5 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012135.F3
also known as 012135-F3
Price (EUR)
-
Quantity:
2 x 2.5 kg
Request bulk or custom format
Chemical Identifiers
CAS544-92-3
IUPAC Nameλ¹-copper(1+) iminomethanide
Molecular FormulaCCuN
InChI KeyDOBRDRYODQBAMW-UHFFFAOYSA-N
SMILES[Cu+].[C-]#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)90% min
Copper(I) cyanide is commonly used as a polymerization catalyst. The compound is useful in organic synthesis, as a catalyst, reagent in the preparation of nitriles(sandmeyer reaction). It is also used in electroplating copper and iron, silver plating, brass plating, and copper-tin alloy plating. It is used to make phthalocyanine dyes and pigments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only