Tantalum carbide, 99.5% (metals basis)
Tantalum carbide, 99.5% (metals basis)
Tantalum carbide, 99.5% (metals basis)
Thermo Scientific Chemicals

Tantalum carbide, 99.5% (metals basis)

CAS: 12070-06-3 | CTa | 192.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 012144.18
also known as 012144-18
Price (EUR)
253,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
253,00
Each
Chemical Identifiers
CAS12070-06-3
IUPAC Namemethanidylidynetantalumylium
Molecular FormulaCTa
InChI KeyDUMHRFXBHXIRTD-UHFFFAOYSA-N
SMILES[C-]#[Ta+]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.5% max.
Tantalum carbides is used in  tungsten carbide. It is also used as a coating for steel moulds in the injection moulding of aluminum alloys. While providing a hard, wear resistant surface, it also provides a low friction mould surface. It is also used in the production of sharp instruments with extreme mechanical resistance and hardness, tool bits for cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only