Nickel(II) nitrate hexahydrate, tech., Ni 19.8% min
Nickel(II) nitrate hexahydrate, tech., Ni 19.8% min
Nickel(II) nitrate hexahydrate, tech., Ni 19.8% min
Thermo Scientific Chemicals

Nickel(II) nitrate hexahydrate, tech., Ni 19.8% min

Nickel plating, manufacturing of ceramic colors | CAS: 13478-00-7 | H12N2NiO12 | 290.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2 kg
Catalog number 012222.36
also known as 012222-36
Price (EUR)
50,50
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
50,50
Each
Chemical Identifiers
CAS13478-00-7
IUPAC Namenickel(2+) hexahydrate dinitrate
Molecular FormulaH12N2NiO12
InChI KeyAOPCKOPZYFFEDA-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ni 19.8% min
Nickel plating, manufacturing of ceramic colorsNickel(II) nitrate hexahydrate is used as a precursor in the manufacturing of other nickel salts and supported nickel catalysts. It finds use as a catalyst in organic synthesis. Further it is used in electroplating and as a mordent in textile dyeing. It is a precursor for preparing nickel sulfide-reduced graphene oxide (RGO) composite powders with high initial capacities and good cycling performance. It can be employed in the preparation of nickel and nickel oxide nanocrystals impregnated O-MWCNTs (oxygen-functionalized multi-walled carbon nanotubes) with potential applications in semiconductor industries. It finds use in improving cycling stability ad rate capability of Li2MnO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only