Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals
Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals
Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cadmium carbonate, >99% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
012229.30
also known as 012229-30
250 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 012229.30
also known as 012229-30
Price (EUR)
106,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS513-78-0
IUPAC Namecadmium(2+) carbonate
Molecular FormulaCCdO3
InChI KeyGKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES[Cd++].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Cadmium carbonate is the raw material for the production of pigments for ceramic and frits. Used as reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only