Vanadium(III) oxide, 95%
Vanadium(III) oxide, 95%
Vanadium(III) oxide, 95%
Thermo Scientific Chemicals

Vanadium(III) oxide, 95%

CAS: 1314-34-7 | O3V2 | 149.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Catalog number 012301.30
also known as 012301-30
Price (EUR)
97,40
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
97,40
Each
Chemical Identifiers
CAS1314-34-7
IUPAC Nameoxo[(oxovanadio)oxy]vanadium
Molecular FormulaO3V2
InChI KeyKFAFTZQGYMGWLU-UHFFFAOYSA-N
SMILESO=[V]O[V]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentIron, Fe: ≤ 0.03%
Impurity contentMolybdenum, Mo: ≤0.01%
Impurity contentPotassium, K: ≤0.01%
Impurity contentSilicon, Si: ≤0.02%
Impurity contentSodium, Na: ≤0.02%
View more
Vanadium(III) oxide catalyzes the direct fluorination reactions. It is used in the coating of carbon nanotubes and as an electrode in lithium ion batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only