Magnesium hydroxide, 95-100.5%
Magnesium hydroxide, 95-100.5%
Magnesium hydroxide, 95-100.5%
Thermo Scientific Chemicals

Magnesium hydroxide, 95-100.5%

Flame Retardant in Plastics, Such As PVC, As It Produces Less Toxic Fumes | CAS: 1309-42-8 | H2MgO2 | 58.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 012367.22
also known as 012367-22
Price (EUR)
33,50
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
33,50
Each
Chemical Identifiers
CAS1309-42-8
IUPAC Namemagnesium(2+) dihydroxide
Molecular FormulaH2MgO2
InChI KeyVTHJTEIRLNZDEV-UHFFFAOYSA-L
SMILES[OH-].[OH-].[Mg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)95-100.5%
It is employed as an additive in certain refractories. It is applied as a non-hazardous alkali in neutralizing acidic wastewater. It is also used in making artificial reefs. It widely used in milk of magnesia antacids and laxatives. Also works as an antiperspirant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only