Iron(III) 2,4-pentanedionate
Iron(III) 2,4-pentanedionate
Iron(III) 2,4-pentanedionate
Thermo Scientific Chemicals

Iron(III) 2,4-pentanedionate

CAS: 14024-18-1 | C15H21FeO6 | 353.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 012534.36
also known as 012534-36
Price (EUR)
156,00
Each
Quantity:
500 g
Request bulk or custom format
Price (EUR)
156,00
Each
Chemical Identifiers
CAS14024-18-1
IUPAC Nameiron(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular FormulaC15H21FeO6
InChI KeyAQBLLJNPHDIAPN-LNTINUHCSA-K
SMILES[Fe+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Volatile content: 0.30%, max
Insoluble material0.30 wt%, max
Appearance (Color)Red
Formpowder
Melting Point170 to 190 deg C

Iron(III) 2,4-pentanedionate is used as a precatalyst and reagent in organic chemistry. It acts as a precursor for highly crystalline (Zn, Fe)Fe2O4 films and magnetic property measurements of these films. It catalyzes the dimerization of isoprene to a mixture of 1,5-dimethyl-1,5-cyclooctadiene and 2,5-dimethyl-1,5-cyclooctadiene, ring-opening polymerization of 1,3-benzoxazine and catalyze the reaction of N-sulfonyl oxaziridines with olefins to form 1,3-oxazolidine products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only