Gold rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)
Thermo Scientific Chemicals

Gold rod, 6.35mm (0.25in) dia, Premion™, 99.99% (metals basis)

CAS: 7440-57-5
Have Questions?
Catalog NumberQuantity
012555.DJ
also known as 012555-DJ
50 mm
Catalog number 012555.DJ
also known as 012555-DJ
Price (EUR)
-
Quantity:
50 mm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS7440-57-5
Percent Purity99.99%
Quantity50 mm
Unit SizeEach