Molybdenum boride, 99% (metals basis)
Molybdenum boride, 99% (metals basis)
Molybdenum boride, 99% (metals basis)
Thermo Scientific Chemicals

Molybdenum boride, 99% (metals basis)

CAS: 12006-98-3 | BMo | 106.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 012563.14
also known as 012563-14
Price (EUR)
84,50
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
84,50
Each
Chemical Identifiers
CAS12006-98-3
IUPAC Namemolybdenioborane
Molecular FormulaBMo
InChI KeyLIGIGBHZRAABEF-UHFFFAOYSA-N
SMILES[B+]=[Mo-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Molybdenum borides have been used as hard coating materials for high temperature structural applications, and also in the form of MoB/CoCr cermet coatings. It is also used as a brazing material especially in electric components. Molybdenum boride has potential industrial applications such as abrasive, corrosion resistant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only