Lithium metaborate, anhydrous, 99.9% (metals basis)
Lithium metaborate, anhydrous, 99.9% (metals basis)
Lithium metaborate, anhydrous, 99.9% (metals basis)
Thermo Scientific Chemicals

Lithium metaborate, anhydrous, 99.9% (metals basis)

CAS: 13453-69-5 | B4H14Li2O7 | 183.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 012591.A1
also known as 012591-A1
Price (EUR)
500,65
Online exclusive
556,00
Save 55,35 (10%)
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
500,65
Online exclusive
556,00
Save 55,35 (10%)
Each
Chemical Identifiers
CAS13453-69-5
IUPAC Nametetraboron heptahydrate dilithium
Molecular FormulaB4H14Li2O7
InChI KeyDPYKRXYVXDYLEY-UHFFFAOYSA-N
SMILES[Li].[Li].[B].[B].[B].[B].O.O.O.O.O.O.O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Used in borate fusion sample preparation of various samples for analysis by XRF, AAS, ICP-MS, ICP-AES and ICP-OES. Used to determine the trace level of elements in contaminated soil. Useful fod dissolving acidic oxides such as SiO2, and Fe2O3. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only