Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v
Thermo Scientific Chemicals

Ruthenium(III) nitrosylnitrate solution, Ru 1.5% w/v

CAS: 34513-98-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Catalog number 012630.AD
also known as 012630-AD
Price (EUR)
121,00
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
121,00
Each
Chemical Identifiers
CAS34513-98-9
IUPAC Nameλ²-ruthenium(2+) iminooxidanium trinitrate
Molecular FormulaN4O10Ru
InChI KeyWOSOOWIGVAKGOC-UHFFFAOYSA-N
SMILES[Ru++].N#[O+].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
FormLiquid
Assay (unspecified)Ruthenium: 1.5% ± 0.225% w/v
Ruthenium(III) nitrosyl nitrate used as a catalyst in various reactions namely hydrogenation, carbonylation, hydrosilylation, hydrofomylation, C-X coupling (eg Heck, Suzuki, Stille) and chiral catalysis reactions. It is also used in the production of thick-film resistors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only