Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%
Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%
Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%
Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%
Thermo Scientific Chemicals

Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate, 98+%

CAS: 50525-27-4 | C30H36Cl2N6O6Ru | 748.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
0.5 g
2 g
Catalog number 012783.02
also known as 012783-02
Price (EUR)
106,00
Each
Add to cart
Quantity:
0.5 g
Request bulk or custom format
Price (EUR)
106,00
Each
Add to cart
Chemical Identifiers
CAS50525-27-4
IUPAC Nameλ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
Molecular FormulaC30H36Cl2N6O6Ru
InChI KeyWHELTKFSBJNBMQ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.1% min
Tris(2,2'-bipyridine)dichlororuthenium(II) hexahydrate is a catalyst used in light-induced redox reactions. It is an important catalyst in studies towards photochemical water splitting. This product can be used as a catalyst in pharmaceutical research. It is widely applied in fields of photoelectrical chemical components, photovoltaic cells, smart biosensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only