Cesium carbonate, 99% (metals basis)
Cesium carbonate, 99% (metals basis)
Cesium carbonate, 99% (metals basis)
Thermo Scientific Chemicals

Cesium carbonate, 99% (metals basis)

Catalyst in ethylene oxide polymerization, in coating spatter-free welding of steel in CO2, and in oxide cathodes | CAS: 534-17-8 | CCs2O3 | 325.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
Catalog number 012887.A1
also known as 012887-A1
Price (EUR)
530,00
Each
Quantity:
1 kg
Request bulk or custom format
Price (EUR)
530,00
Each
Chemical Identifiers
CAS534-17-8
IUPAC Namedicaesium(1+) carbonate
Molecular FormulaCCs2O3
InChI KeyFJDQFPXHSGXQBY-UHFFFAOYSA-L
SMILES[Cs+].[Cs+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Cesium carbonate is the precursor for other cesium compounds. It acts as a base in sensitive organic reactions. Suzuki, Heck and Sonogashira synthesis are some of the important reactions involving cesium carbonate. It finds use in solar cells as it increases the power conversion efficiency of cells through the transfer of electrons. It is also used in the production of special optical glasses, petroleum catalytic additives, special ceramics, and in sulfuric acid industry. It is useful in the N-alkylation (of sulfonamides, beta-lactams, indoles, heterocycles, and several sensitive nitrogen compounds), carbamination of amines, carbonylation of alcohols, and aerobic oxidation of alcohols into carbonyl compounds without over oxidations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only