Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia, ISA Type R Standard Grade Thermocouple, Thermo Scientific Chemicals

For ISA Type R Standard Grade Thermocouple
Have Questions?
Catalog NumberQuantity
013018.BY
also known as 013018-BY
100 cm
Catalog number 013018.BY
also known as 013018-BY
Price (EUR)
-
Quantity:
100 cm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 87:13 wt%
Chemical Name or MaterialPlatinum 13 wt% Rhodium wire
Density19.595G/CC
MDL NumberMFCD00198186
Name Note0.5mm (0.02 in.) dia., Positive leg Type R
View more

Platinum 13 wt% Rhodium wire, 0.5mm (0.02in) dia is used as thermocouples and heat flux gauges. It can be used for high temperature measurements because high cost and low sensitivity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only