Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis)
Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis)
Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis)
Thermo Scientific Chemicals

Bismuth germanium oxide, Puratronic™, 99.9995% (metals basis)

CAS: 12233-56-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013029.14
also known as 013029-14
Price (EUR)
154,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
154,00
Each
Chemical Identifiers
CAS12233-56-6
IUPAC Nametetrabismuth trigermanium dodecahydrate
Molecular FormulaBi4Ge3H24O12
InChI KeyORCSMBGZHYTXOV-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Ge].[Ge].[Ge].[Bi].[Bi].[Bi].[Bi]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.0005% max.
Bismuth germanium oxide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only