Niobium(IV) selenide, 99.8% (metals basis)
Niobium(IV) selenide, 99.8% (metals basis)
Niobium(IV) selenide, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium(IV) selenide, 99.8% (metals basis)

CAS: 12034-77-4 | NbSe2 | 250.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number 013101.09
also known as 013101-09
Price (EUR)
98,10
Each
Quantity:
10 g
Request bulk or custom format
Price (EUR)
98,10
Each
Chemical Identifiers
CAS12034-77-4
IUPAC Namediselanylideneniobium
Molecular FormulaNbSe2
InChI KeyCXRFFSKFQFGBOT-UHFFFAOYSA-N
SMILES[Se]=[Nb]=[Se]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.

It is used as a potential substance for thin film solar cells as back contact material. It is widely used in electricity brush & special bearings because of its conductivity and lubricity property.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only