Antimony(III) selenide, 99.999% (metals basis)
Antimony(III) selenide, 99.999% (metals basis)
Antimony(III) selenide, 99.999% (metals basis)
Thermo Scientific Chemicals

Antimony(III) selenide, 99.999% (metals basis)

CAS: 1315-05-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013130.06
also known as 013130-06
Price (EUR)
204,00
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
204,00
Each
Chemical Identifiers
CAS1315-05-5
IUPAC Namebicyclo[1.1.1]distibaselenane
Molecular FormulaSb2Se3
InChI KeyGNZJTRGEKSBAAS-UHFFFAOYSA-N
SMILES[Se]1[Sb]2[Se][Sb]1[Se]2
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.001% max.
It is used to prepare large zintl ion, it is now actively explored for thin film solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only