Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO)
Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO)
Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Lanthanum(III) bromide hydrate, REacton™, 99.99% (REO)

CAS: 224183-16-8 | Br3La | 378.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 013198.30
also known as 013198-30
Price (EUR)
577,00
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
577,00
Each
Chemical Identifiers
CAS224183-16-8
IUPAC Namelanthanum(3+) tribromide
Molecular FormulaBr3La
InChI KeyXKUYOJZZLGFZTC-UHFFFAOYSA-K
SMILES[Br-].[Br-].[Br-].[La+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
Lanthanum(III) bromide is used as a scintillation material in certain applications. It is used in gamma spectroscopy radioisotope detection. It is used as a source of lanthanum in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only