Europium(III) carbonate hydrate, REacton™, 99.99% (REO)
Europium(III) carbonate hydrate, REacton™, 99.99% (REO)
Europium(III) carbonate hydrate, REacton™, 99.99% (REO)
Thermo Scientific Chemicals

Europium(III) carbonate hydrate, REacton™, 99.99% (REO)

CAS: 86546-99-8 | C3Eu2O9 | 483.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number 013212.14
also known as 013212-14
Price (EUR)
987,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
987,00
Each
Chemical Identifiers
CAS86546-99-8
IUPAC Namedieuropium(3+) tricarbonate
Molecular FormulaC3Eu2O9
InChI KeyMBFNJTXDYDGXDL-UHFFFAOYSA-H
SMILES[Eu+3].[Eu+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.01% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only