Lithium tetraborate, 99% (metals basis)
Lithium tetraborate, 99% (metals basis)
Lithium tetraborate, 99% (metals basis)
Lithium tetraborate, 99% (metals basis)
Thermo Scientific Chemicals

Lithium tetraborate, 99% (metals basis)

CAS: 12007-60-2 | B4Li2O7 | 169.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 013419.18
also known as 013419-18
Price (EUR)
42,90
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
42,90
Each
Chemical Identifiers
CAS12007-60-2
IUPAC Namedilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular FormulaB4Li2O7
InChI KeyRDZCISUGRQUANA-UHFFFAOYSA-N
SMILES[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities1% max.
Used in the laboratory as LB (Lithium borate) buffer for gel electrophoresis of DNA and RNA. Employed in the borax fusion method to modify mineral powder samples for analysis by WDXRF (Wavelength Dispersive X-Ray Fluorescence) Spectroscopy. Acts as an ingredient in the manufacture of glasses and ceramics. Useful fod dissolving basic oxides such as CaO and MgO. A mixture of lithium metaborate and lithium tetraborate is useful for dissolving most of the inorganic oxides to enable spectrochemical analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only