Lithium oxalate, 99+%
Lithium oxalate, 99+%
Lithium oxalate, 99+%
Thermo Scientific Chemicals

Lithium oxalate, 99+%

CAS: 553-91-3 | C2Li2O4 | 101.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Catalog number 013426.18
also known as 013426-18
Price (EUR)
45,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
45,00
Each
Chemical Identifiers
CAS553-91-3
IUPAC Namedilithium(1+) oxalate
Molecular FormulaC2Li2O4
InChI KeyYNQRWVCLAIUHHI-UHFFFAOYSA-L
SMILES[Li+].[Li+].[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)White to off-white
Assay from Supplier's CofA≥99.0%
Lithium oxalate is used as lithium battery electrolyte. It is also used as an analytical reagent and reducing reagent. Further, it is used as an intermediate in the manufacture of other chemicals such as pharmaceuticals, agrochemicals, dyestuffs and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only