Dysprosium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Dysprosium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Dysprosium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
Thermo Scientific Chemicals

Dysprosium(III) fluoride, anhydrous, REacton™, 99.9% (REO)

CAS: 13569-80-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 013656.14
also known as 013656-14
Price (EUR)
108,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
108,00
Each
Chemical Identifiers
CAS13569-80-7
IUPAC Namedysprosium(3+) trifluoride
Molecular FormulaDyF3
InChI KeyFWQVINSGEXZQHB-UHFFFAOYSA-K
SMILES[F-].[F-].[F-].[Dy+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
Dysprosium(III) fluoride, anhydrous is used for vacuum deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only