Cerium, plasma standard solution, Specpure™ Ce 1000μg/mL
Cerium, plasma standard solution, Specpure™ Ce 1000μg/mL
Cerium, plasma standard solution, Specpure™ Ce 1000μg/mL
Thermo Scientific Chemicals

Cerium, plasma standard solution, Specpure™ Ce 1000μg/mL

CAS: 7440-45-1 | Ce | 140.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
Catalog number 013815.AE
also known as 013815-AE
Price (EUR)
73,65
Online Exclusive
82,00
Save 8,35 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
73,65
Online Exclusive
82,00
Save 8,35 (10%)
Each
Specifications
CAS7440-45-1
CAS7732-18-5
ChEBICHEBI:33369
Chemical Name or MaterialCerium
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only