Sodium, plasma standard solution, Specpure™ Na 1000μg/mL
Thermo Scientific Chemicals

Sodium, plasma standard solution, Specpure™ Na 1000μg/mL

Na2CO3 in 5% HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013832.AD
also known as 013832-AD
Price (EUR)
55,60
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
55,60
Each
Specifications
DOT InformationHazard Class: 8; Packaging Group: III
EINECS Number231-714-2
Health Hazard 1H290-H314-H335
Health Hazard 3P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
MDL NumberMFCD00085307
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only