Zinc, plasma standard solution, Specpure™ Zn 1000μg/mL
Zinc, plasma standard solution, Specpure™ Zn 1000μg/mL
Zinc, plasma standard solution, Specpure™ Zn 1000μg/mL
Thermo Scientific Chemicals

Zinc, plasma standard solution, Specpure™ Zn 1000μg/mL

CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013835.AD
also known as 013835-AD
Price (EUR)
55,00
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
55,00
Each
Specifications
CAS7440-66-6
CAS7732-18-5
ChEBICHEBI:30185
Chemical Name or MaterialZinc
ColorColorless
View more
Zinc, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only