Selenium, plasma standard solution, Specpure™ Se 1000μg/mL
Selenium, plasma standard solution, Specpure™ Se 1000μg/mL
Selenium, plasma standard solution, Specpure™ Se 1000μg/mL
Thermo Scientific Chemicals

Selenium, plasma standard solution, Specpure™ Se 1000μg/mL

Se 1000μg/mL | CAS: 7697-37-2 | HNO3 | 63.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013845.AD
also known as 013845-AD
Price (EUR)
53,60
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
53,60
Each
Specifications
CAS7697-37-2
ChEBICHEBI:48107
Chemical Name or MaterialSelenium
Concentration or Composition (by Analyte or Components)Nitric acid: 5%, Selenium: 0.15%, Water: 94.9%
DOT InformationDOT Class: 8, Packaging Group: III
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only