Tin, plasma standard solution, Specpure™ Sn 1000μg/mL
Tin, plasma standard solution, Specpure™ Sn 1000μg/mL
Tin, plasma standard solution, Specpure™ Sn 1000μg/mL
Thermo Scientific Chemicals

Tin, plasma standard solution, Specpure™ Sn 1000μg/mL

CAS: 7440-31-5 | Sn | 118.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013863.AE
also known as 013863-AE
Price (EUR)
82,65
キャンペーン価格
91,80
Save 9,15 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
82,65
キャンペーン価格
91,80
Save 9,15 (10%)
Each
Specifications
CAS7440-31-5
CAS7647-01-0
CAS7732-18-5
ChEBICHEBI:32990
Chemical Name or MaterialTin
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only