Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL
Thermo Scientific Chemicals

Lanthanum, plasma standard solution, Specpure™ La 1000μg/mL

CAS: 1312-81-8 | La2O3 in 5% HNO3
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
500 mL
Catalog number 013870.AD
also known as 013870-AD
Price (EUR)
53,20
Each
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
53,20
Each
Specifications
CAS1312-81-8
CAS7697-37-2
CAS7732-18-5
Chemical Name or MaterialLanthanum
ColorColorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only