Cyclopentadienylthallium
Cyclopentadienylthallium
Cyclopentadienylthallium
Thermo Scientific Chemicals

Cyclopentadienylthallium

CAS: 34822-90-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 013958.06
also known as 013958-06
Price (EUR)
85,10
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
85,10
Each
Chemical Identifiers
CAS34822-90-7
IUPAC Name(cyclopenta-2,4-dien-1-yl)thallane
Molecular FormulaC5H5Tl
InChI KeyRKLJDPOVAWGBSD-UHFFFAOYSA-N
SMILES[Tl]C1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale brown to brown
FormPowder
Elemental AnalysisC: ≥20.9%
Elemental AnalysisH: ≥1.7%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only